missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford SDS Pellets
Description
New micro-pellets allow convenient weighing and handling without the mess and odor of? powders.
New micro-pellets allow convenient weighing and handling without the mess and odor of powders. High purity SDS is additive free. Easy to dissolve in water for denaturing proteins prior to gel elecrophoresis.
Specifications
Specifications
| Chemical Name or Material | SDS, Pellets |
| CAS | 151-21-3 |
| Quantity | 1 kg |
| Molecular Formula | C12H25NaO4S |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?