missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford SDS Pellets
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Description
New micro-pellets allow convenient weighing and handling without the mess and odor of? powders.
New micro-pellets allow convenient weighing and handling without the mess and odor of powders. High purity SDS is additive free. Easy to dissolve in water for denaturing proteins prior to gel elecrophoresis.
Specifications
Specifications
| Chemical Name or Material | SDS, Pellets |
| CAS | 151-21-3 |
| Quantity | 100 g |
| Molecular Formula | C12H25NaO4S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction