missing translation for 'onlineSavingsMsg'
Learn More

SDS-PAGE Buffer, 5X Powder, Electrophoresis, Fisher BioReagents

Product Code. 10618203
Change view
Click to view available options
Quantity:
92 g
Unit Size:
92g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10618203 92 g 92g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10618203 Supplier Fisher BioReagents Supplier No. BP139892
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Mix of tris-glycine-SDS (TGS) for electrophoresis buffer
  • Tris-glycine-SDS is the most commonly used running buffer for SDS-PAGE
  • Pre-weighed and pre-mixed ingredients to make 1L of 5X tris-glycine-SDS buffer
  • Eliminates the need to weigh ingredients - simply dissolve powder in pure water
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Glycine-SDS
CAS 56-40-6, 77-86-1, 151-21-3
Color White
pH 8.3
Physical Form Solid
Quantity 92 g
Packaging Amber glass bottle
Synonym TGS Buffer
Grade Electrophoresis
compliance-icons
Product Identifier
  • SDS-PAGE-Buffer Mix
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.