missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDOS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107200
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
NUDT16L1 is a probable adapter protein, which may link syndecan-4 (SDC4) and paxilin (TGFB1I1 and PXN) receptors.
Specifications
| SDOS | |
| Polyclonal | |
| Unconjugated | |
| NUDT16L1 | |
| nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1; nudix hydrolase 16 like 1; NUDT16L1; NUDT16-like protein 1; Protein syndesmos; SDOS; syndesmos; TIRR; Tudor-interacting repair regulator protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84309 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9BRJ7 | |
| NUDT16L1 | |
| Synthetic peptide directed towards the middle region of human NUDT16L1 (aa 78-128). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction