missing translation for 'onlineSavingsMsg'
Learn More

SDNSF/MCFD2 Antibody (294301) [DyLight 405], Novus Biologicals™

Product Code. 30732665 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732665 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732665 Supplier Novus Biologicals Supplier No. FAB2357E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SDNSF/MCFD2 Monoclonal antibody specifically detects SDNSF/MCFD2 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDNSF/MCFD2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 294301
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp686G21263, F5F8D2, LMAN1IP, multiple coagulation factor deficiency 2, multiple coagulation factor deficiency protein 2, neural stem cell derived neuronal survival protein, Neural stem cell-derived neuronal survival protein, SDNSFF5F8D
Host Species Mouse
Immunogen E. coli-derived recombinant human SDNSF/MCFD2, Glu27-Gln146, Accession # Q8NI22
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 90411
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.