missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDK1 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 79%; Dog: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Rabbit: 93%; Zebrafish: 79%.
The protein encoded by this gene is a member of the immunoglobulin superfamily. The protein contains six immunoglobulin-like domains and thirteen fibronectin type III domains. Fibronectin type III domains are present in both extracellular and intracellular proteins and tandem repeats are known to contain binding sites for DNA, heparin and the cell surface. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | SDK1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | SDK1 |
| Gene Accession No. | Q7Z5N4 |
| Gene Alias | 5330440E10; 6720466O15Rik; Protein sidekick-1; RGD1560686; Sdk1; sidekick cell adhesion molecule 1; sidekick homolog 1; sidekick homolog 1 (chicken); sidekick homolog 1, cell adhesion molecule |
| Gene Symbols | SDK1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?