missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDHD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561081
This item is not returnable.
View return policy
Description
Immunogen sequence: RTPVVRPAHI SAFLQDRPIP EWCGVQHIHL SPSHHSGSKA ASLHW Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.
The mitochondrial succinate dehydrogenase complex subunit D (SDHD) is one of four proteins that make up the tricarboxylic cycle enzyme succinate dehydrogenase (SCH). Studies have shown that mutations in SDHD often leads to hereditary paragangliomas, usually benign tumors of the autonomic nervous system, suggesting that SDHD also plays a role as a tumor-suppressor gene. In one family with a nonsense mutation (R22X) in the SDHD gene, a loss of heterozygosity was found in the paragangliomas, and within these tumors the enzymatic activity of Complex II in the mitochondrial respiratory chain was completely abolished. Furthermore, high levels of angiogenic factors EPAS1 and VEGF was observed, which may stimulate tumor growth.
Specifications
| SDHD | |
| Polyclonal | |
| Unconjugated | |
| SDHD | |
| 3110001M13Rik; AVLL5809; C78570; CBT1; CII-4; CWS3; CybS; PGL; PGL1; PRO19626; QPs3; SDH4; SDHD; Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial; succinate dehydrogenase complex subunit D; succinate dehydrogenase complex subunit D integral membrane protein; succinate dehydrogenase complex, subunit D, integral membrane protein; succinate-ubiquinone oxidoreductase cytochrome b small subunit; Succinate-ubiquinone reductase membrane anchor subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6392 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14521 | |
| SDHD | |
| Recombinant protein corresponding to Human SDHD. Recombinant protein control fragment (Product #RP-100470). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction