missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHD Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93065-0.02ml
This item is not returnable.
View return policy
Description
SDHD Polyclonal antibody specifically detects SDHD in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SDHD | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CBT1, CII-4, CybS, PGL, PGL1, QPs3, SDH4, succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial, Succinate dehydrogenase complex subunit D, succinate dehydrogenase complex, subunit D, integral membrane protein, succinate dehydrogenase ubiquinone cytochrome B small subunit, Succinate-ubiquinone oxidoreductase cytochrome b small subunit, Succinate-ubiquinone reductase membrane anchor subunit | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human SDHD (NP_002993.1). HLSPSHHSGSKAASLHWTSERVVSVLLLGLLPAAYLNPCSAMDYSLAAALTLHGHWGLGQVVTDYVHGDALQKAAKAGLLALSALTFAGLCYFNYHDVGIC | |
| 0.02 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6392 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction