missing translation for 'onlineSavingsMsg'
Learn More

SDHD Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18407355 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407355 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407355 Supplier Novus Biologicals Supplier No. NBP281948AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SDHD Polyclonal antibody specifically detects SDHD in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDHD
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CBT1, CII-4, CybS, PGL, PGL1, QPs3, SDH4, succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial, Succinate dehydrogenase complex subunit D, succinate dehydrogenase complex, subunit D, integral membrane protein, succinate dehydrogenase ubiquinone cytochrome B small subunit, Succinate-ubiquinone oxidoreductase cytochrome b small subunit, Succinate-ubiquinone reductase membrane anchor subunit
Host Species Rabbit
Immunogen SDHD antibody was raised against a 15 amino acid synthetic peptide near the center of human SDHD. The immunogen is located within amino acids 30 - 80 of SDHD. Amino Acid Squence: HLSPSHHSGSKAASL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6392
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.