Learn More
Invitrogen™ SDHB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588051
Description
Immunogen sequence: AQTAAATAPR IKKFAIYRWD PDKAGDKPHM QTYEVDLNKC GPMVLDALIK IKNEVDSTLT FRRSCREGIC GSCAMNINGG NTLACTRRID TNLNKVSKIY PLPHMYVIKD LVPDLSNFYA QYKSIEPYLK KKDESQEGKQ QYLQSIEERE KLDGLYECIL CACCSTSCPS YWWNGDKYLG PAVLMQAYRW MIDSRDDFTE ERLAKLQDPF SLYRCHTIMN CTRTCPKGLN PGKAIAEIKK MMATYKEKKA SV; Positive Samples: HepG2, HeLa, U-87MG, Mouse brain, Mouse liver, Mouse kidney, Mouse heart, Rat liver, Rat brain; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.
Specifications
| SDHB | |
| Polyclonal | |
| Unconjugated | |
| Sdhb | |
| 0710008N11Rik; CWS2; fj46d06; FLJ92337; I79_022285; Ip; iron-sulfur subunit; iron-sulfur subunit of complex II; OTTHUMP00000044624; PGL4; SDH; SDH1; SDH2; Sdhb; SDHB protein; SDHB; sdhB; SDHIP; succinate dehydrogenase [ubiquinone] iron-sulfur subunit; Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial; succinate dehydrogenase complex iron sulfur subunit B; succinate dehydrogenase complex subunit B; succinate dehydrogenase complex subunit B, iron sulfur (Ip); succinate dehydrogenase complex, subunit B, iron sulfur (Ip); succinate dehydrogenase Ip subunit; succinate dehydrogenase iron-protein subunit; wu:fj46d06 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 298596, 6390, 67680 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.82 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P21912, P21913, Q9CQA3 | |
| Sdhb | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-280 of human SDHB (NP_0029912). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.