Learn More
SDHB Mouse anti-Human, Unconjugated, Clone: 1A2A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67600-1-IG-20UL
 View more versions of this product
Product Code. 16880316
  • £135.00 / 20µL
Description
The SDHB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SDHB. This antibody recognizes human, mouse, rat antigen. The SDHB antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SDHB
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1A2A5
Unconjugated
20 μL
6390
IgG1