missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SDHB Polyclonal specifically detects SDHB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | SDHB |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SDHB. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?