missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDHA Monoclonal Antibody (D1), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551137
This item is not returnable.
View return policy
Description
This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.
Specifications
| SDHA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q8K2B3, Q920L2 | |
| SDHA | |
| Recombinant Mouse SDHA protein, Val4-Ile300 (Accession #Q8K2B3). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2a κ |
| Western Blot | |
| D1 | |
| Unconjugated | |
| SDHA | |
| 1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; Flavoprotein subunit of complex II; FP; im:7141001; mitochondrial succinate dehydrogenase complex subunit A; PGL5; SDH1; SDH2; SDHA; SDHA protein; SDHF; SDHFP1; si:dkeyp-84f11.2; succinate dehydrogenase [ubiquinone] flavoprotein subunit; succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex flavoprotein subunit A; succinate dehydrogenase complex subunit A; succinate dehydrogenase complex subunit A flavoprotein; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp); succinate dehydrogenase flavoprotein subunit; succinate dehydrogenase flavoprotein subunit A; zgc:56051 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 157074, 66945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction