missing translation for 'onlineSavingsMsg'
Learn More

SDHA Monoclonal Antibody (2E3GC12FB2AE2), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen 459200

Code : 50

Additional Details : Weight : 0.50000kg

Product Code. 10771814

  • £466.00 / Each
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

ICC application requires heat-induced antigen retrieval where aldehydes are used as fixatives. Use 20min incubation at 90-100°C in 0.1 M Tris/HCl pH 9.5 with 5% urea (wt/vol). Positive control: Human heart mitochondria.

SDHA Monoclonal antibody specifically detects SDHA in Bovine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.
Specifications

Specifications

SDHA
Monoclonal
1 mg/mL
HEPES buffered saline with 0.02% sodium azide; pH 7.5
P31039, P31040, Q8K2B3, Q920L2
SDHA
Full length native protein (purified) corresponding to Cow SDHA. Purified mitochondrial complex II (Cow).
100 μg
Primary
Bovine, Human, Mouse, Rat
Antibody
IgG1 κ
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
2E3GC12FB2AE2
Unconjugated
SDHA
1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; Flavoprotein subunit of complex II; FP; im:7141001; mitochondrial succinate dehydrogenase complex subunit A; PGL5; SDH1; SDH2; SDHA; SDHA protein; SDHF; SDHFP1; si:dkeyp-84f11.2; succinate dehydrogenase [ubiquinone] flavoprotein subunit; succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex flavoprotein subunit A; succinate dehydrogenase complex subunit A; succinate dehydrogenase complex subunit A flavoprotein; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp); succinate dehydrogenase flavoprotein subunit; succinate dehydrogenase flavoprotein subunit A; zgc:56051
Mouse
IgG fraction
RUO
157074, 281480, 6389, 66945
4° C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers