missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10517-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a stromal cell derived factor that is a member of the CREC protein family. The encoded protein contains six EF-hand motifs and calcium-binding motifs. This protein localizes to the Golgi lumen and may be involved in regulating calcium dependent cellular activities.Specifications
| SDF4 | |
| Polyclonal | |
| Unconjugated | |
| SDF4 | |
| 45 kDa calcium binding protein, Cab45, SDF 4, SDF4, stromal cell derived factor 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 155173, 20318, 51150 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q61112, Q91ZS3, Q9BRK5 | |
| SDF4 | |
| SDF4 Fusion Protein Ag0806 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction