missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDF4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15634715
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634715 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15634715 Supplier Invitrogen™ Supplier No. PA550785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SDF4 protein.

This gene encodes a stromal cell derived factor that is a member of the CREC protein family. The encoded protein contains six EF-hand motifs and calcium-binding motifs. This protein localizes to the Golgi lumen and may be involved in regulating calcium dependent cellular activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDF4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SDF4
Gene Accession No. Q9BRK5
Gene Alias 45 kDa calcium-binding protein; cab45; PSEC0034; Sdf4; SDF-4; stromal cell derived factor 4; stromal cell-derived factor 4
Gene Symbols SDF4
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human stromal cell derived factor 4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51150
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.