missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | SDF4 |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228958
|
Novus Biologicals
NBP3-35551-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232831
|
Novus Biologicals
NBP3-35551-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SDF4 Polyclonal antibody specifically detects SDF4 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| SDF4 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| 45 kDa calcium-binding protein, CAB45, Cab45Stromal cell-derived factor 4, calcium binding protein, SDF-4, stromal cell derived factor 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 37-200 of human SDF4 (NP_057260.2).,, Sequence:, RPANHSSTRERVANREENEILPPDHLNGVKLEMDGHLNRGFHQEVFLGKDLGGFDEDAEPRRSRRKLMVIFSKVDVNTDRKISAKEMQRWIMEKTAEHFQEAMEESKTHFRAVDPDGDGHVSWDEYKVKFLASKGHSEKEVADAIRLNEELKVDEETQEVLENL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 51150 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title