missing translation for 'onlineSavingsMsg'
Learn More

SDE2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18417804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417804 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417804 Supplier Novus Biologicals Supplier No. NBP119139JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SDE2 Polyclonal antibody specifically detects SDE2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SDE2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C1orf55, chromosome 1 open reading frame 55, dJ671D7.1, FLJ35382, hypothetical protein LOC163859, RP4-671D7.1, SDE2 telomere maintenance homolog (S. pombe)
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human chromosome 1 open reading frame 55 using the numbering given in entry NP_689821.3 (GeneID 163859).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163859
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.