missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDCCAG8 Rabbit anti-Canine, Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13471-1-AP-150UL
This item is not returnable.
View return policy
Description
13471-1-AP can recognize all four isoforms.
SDCCAG8 (serologically defined colon cancer antigen 8), also known as CCCAP (centrosomal colon cancer autoantigen protein), HSPC085 or NY-CO-8, is a 713 amino acid cytoplasmic protein that is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa and renal cancer tumors. Existing as a homodimer, SDCCAG8 localizes to centrioles and interacts with oral-facial-digital syndrome 1 (ODF1), which is associated with nephronophthisis-related ciliopathies (NPHP-RC), a recessive disorder that is characterized by dysplasia or degeneration of the kidney, retina and cerebellum. SDCCAG8 exists as four alternatively spliced isoforms and is encoded by a gene located on humanc chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.Specifications
| SDCCAG8 | |
| Polyclonal | |
| Unconjugated | |
| SDCCAG8 | |
| Antigen NY CO 8, BBS16, CCCAP, hCCCAP, HSPC085, NY CO 8, SDCCAG8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10806, 305002, 490374, 556980, 76816 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B8JK76, Q80UF4, Q86SQ7 | |
| sdccag8 | |
| SDCCAG8 Fusion Protein Ag4264 | |
| 150 μL | |
| Primary | |
| Canine, Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction