missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDCCAG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£470.00
Specifications
| Antigen | SDCCAG8 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SDCCAG8 Polyclonal specifically detects SDCCAG8 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| SDCCAG8 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CCCAPAntigen NY-CO-8, Centrosomal colon cancer autoantigen protein, hCCCAP, NPHP10, NY-CO-8, serologically defined colon cancer antigen 8, SLSN7 | |
| SDCCAG8 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 10806 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LIQCDQLRKELERQAERLEKELASQQEKRAIEKDMMKKEITKEREYMGSKMLILSQNIAQLEAQVEKVTKEKISAINQLEEIQSQLASREMDVTKV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title