missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDCCAG8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56699
This item is not returnable.
View return policy
Description
SDCCAG8 Polyclonal specifically detects SDCCAG8 in Human samples. It is validated for Western Blot.
Specifications
| SDCCAG8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CCCAPAntigen NY-CO-8, Centrosomal colon cancer autoantigen protein, hCCCAP, NPHP10, NY-CO-8, serologically defined colon cancer antigen 8, SLSN7 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10806 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q86SQ7 | |
| SDCCAG8 | |
| Synthetic peptides corresponding to SDCCAG8(serologically defined colon cancer antigen 8) The peptide sequence was selected from the middle region of SDCCAG8. Peptide sequence IQCDQLRKELERQAERLEKELASQQEKRAIEKDMMKKEITKEREYMGSKM. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Bovine: 92%; Guinea pig: 92%; Rabbit: 92%; Rat: 92%; Canine: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction