missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDCBP, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00006386-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MSLYPSLEDLKVDKVIQAQTAFSANPANPAILSEASAPIPHDGNLYPRLYPELSQYMGLSLNEEEIRANVAVVSGAPLQGQLVARPSSINYMVAPVTGNDVGIRRAEIKQGIREVILCKDQDGKIGLRLKSIDNGIFVQLVQANSPASLVGLRFGDQVLQINGENCAGWSSDKAHKVLKQAFGEKITMTIRDRPFERTITMHKDSTGHVGFIFKNGKITSIVKDSSAARNGLLTEHNICEINGQNVIGLKDSQIADILSTSGTVVTITIMPAFIFEHIIKRMAPSIMKSLMDHTIPEVSpecifications
| SDCBP | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_001007067 | |
| SDCBP | |
| SDCBP (NP_001007068.1, 1 a.a. ∼ 298 a.a) full-length human protein. | |
| RUO | |
| 6386 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| syndecan binding protein (syntenin) | |
| MDA-9/ST1/SYCL/TACIP18 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Safety and Handling
Product Identifier
Supplemental information
- SDCBP purified MaxPab mouse polyclonal antibody (B01P)
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate, potassium phosphate, sodium chloride, potassium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction