missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDAD1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98637V
This item is not returnable.
View return policy
Description
SDAD1 Polyclonal specifically detects SDAD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SDAD1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| FLJ10498, SDA1 domain containing 1 | |
| Produced in rabbits immunized with E. coli-derived Human SDAD1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 55153 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction