missing translation for 'onlineSavingsMsg'
Learn More

SD 0006, MedChemExpress

Product Code. p-200099659
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480707

Brand: Medchem Express HY11087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SD 0006 (SD-06) is an orally active, selective, ATP-competitive and potent diaryl pyrazole inhibitor of p38α MAP kinase, with an IC50 of 110 nM for p38α.

SD 0006 (SD-06) is an orally active, selective, ATP-competitive and potent diaryl pyrazole inhibitor of p38α MAP kinase, with an IC50 of 110 nM for p38α.

Specifications

Chemical Name or Material SD 0006
Physical Form Solid
CAS 271576-80-8
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20ClN5O2
For Use With (Application) COVID-19-immunoregulation
Synonym SD-06
Solubility Information DMSO : 50 mg/mL (125.67 mM; Need ultrasonic)
SMILES OCC(N1CCC(CC1)C2=NNC(C3=CC=C(C=C3)Cl)=C2C4=NC=NC=C4)=O
Molecular Weight (g/mol) 397.86
Formula Weight 397.86
Percent Purity 98.6%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.