missing translation for 'onlineSavingsMsg'
Learn More

SD 0006, MedChemExpress

Product Code. 16480707
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480707 50 mg 50mg
16474388 10 mg 10mg
16455286 100 mg 100mg
16407117 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480707 Supplier Medchem Express Supplier No. HY11087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SD 0006 (SD-06) is an orally active, selective, ATP-competitive and potent diaryl pyrazole inhibitor of p38α MAP kinase, with an IC50 of 110 nM for p38α.

SD 0006 (SD-06) is an orally active, selective, ATP-competitive and potent diaryl pyrazole inhibitor of p38α MAP kinase, with an IC50 of 110 nM for p38α.

Specifications

Chemical Name or Material SD 0006
Physical Form Solid
CAS 271576-80-8
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H20ClN5O2
For Use With (Application) COVID-19-immunoregulation
Synonym SD-06
Solubility Information DMSO : 50 mg/mL (125.67 mM; Need ultrasonic)
SMILES OCC(N1CCC(CC1)C2=NNC(C3=CC=C(C=C3)Cl)=C2C4=NC=NC=C4)=O
Molecular Weight (g/mol) 397.86
Formula Weight 397.86
Percent Purity 98.6%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.