missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCYL1BP1 Antibody (OTI4F11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74029CL1
This item is not returnable.
View return policy
Description
SCYL1BP1 Monoclonal antibody specifically detects SCYL1BP1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| SCYL1BP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ11752, golgin, RAB6-interacting, GONTKL-binding protein 1, hNTKL-BP1, MGC51263, MGC70512, N-terminal kinase-like-binding protein 1, NTKL-BP1, NTKLBP1SCY1-like 1-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCYL1-BP1, SCYL1BP1SCYL1-binding protein 1 | |
| Human recombinant protein fragment corresponding to amino acids 35-394 of human GORAB(NP_689494) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI4F11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 92344 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction