missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCYL1BP1 Polyclonal antibody specifically detects SCYL1BP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | SCYL1BP1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ11752, golgin, RAB6-interacting, GONTKL-binding protein 1, hNTKL-BP1, MGC51263, MGC70512, N-terminal kinase-like-binding protein 1, NTKL-BP1, NTKLBP1SCY1-like 1-binding protein 1, RAB6-interacting golgin, SCY1-like 1 binding protein 1, SCYL1-BP1, SCYL1BP1SCYL1-binding protein 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human SCYL1BP1 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?