missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCYL1 Polyclonal Antibody

SCYL1 Polyclonal Antibody
Brand: Invitrogen PA593185
This item is not returnable.
View return policy
Description
SCYL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| SCYL1 | |
| Polyclonal | |
| Unconjugated | |
| Scyl1 | |
| 105 kDa kinase-like protein; 2810011O19Rik; C85140; Coated vesicle-associated kinase of 90 kDa; CVAK90; GKLP; HT019; likely ortholog of mouse N-terminal kinase-like protein; mdf; mfd; Mitosis-associated kinase-like protein NTKL; NKTL; N-terminal kinase-like protein; NTKL; P105; SCAR21; SCY1 like pseudokinase 1; SCY1-like 1 (S. cerevisiae); SCY1-like 1 splice variant 13; SCY1-like 1 splice variant 15; SCY1-like protein 1; SCY1-like, kinase-like 1; Scyl1; TAPK; TEIF; Telomerase regulation-associated protein; Telomerase transcriptional element-interacting factor; telomerase transcriptional elements-interacting factor; teratoma-associated tyrosine kinase; TRAP; ubiquitous protein kinase-like (105 kDa) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 293684, 57410, 78891 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.92 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5M9F8, Q96KG9, Q9EQC5 | |
| Scyl1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 460-740 of human SCYL1 (NP_0657313). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction