missing translation for 'onlineSavingsMsg'
Learn More

SCX Mouse anti-Human, Clone: PCRP-SCXA-2D11, Novus Biologicals™

Mouse Monoclonal Antibody

£399.00

Specifications

Antigen SCX
Clone PCRP-SCXA-2D11
Concentration 0.2 mg/mL
Dilution Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18634459
View Documents
Novus Biologicals
NBP3-07598-100ug
100 μg
£399.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SCX Monoclonal specifically detects SCX in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications

Specifications

SCX
0.2 mg/mL
Flow Cytometry, Immunofluorescence, Peptide Array
Unconjugated
Mouse
Human
BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB
Recombinant full-length human SCX protein
Primary
Store at 4°C.
PCRP-SCXA-2D11
Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
642658
IgG2a
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.