missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCX Antibody (PCRP-SCXA-1D2), Alexa Fluor™ 405, Novus Biologicals™
Description
SCX Monoclonal antibody specifically detects SCX in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
Specifications
| Antigen | SCX |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | PCRP-SCXA-1D2 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BHLHA41, BHLHa48, Class A Basic Helix-Loop-Helix Protein 41, Class A Basic Helix-Loop-Helix Protein 48, Class II BHLH Protein Scleraxis, Scleraxis Basic Helix-Loop-Helix Transcription Factor, Scleraxis BHLH Transcription Factor, Scleraxis Homolog (Mouse), Scleraxis Homolog A, Scleraxis Homolog A (Mouse), Scleraxis Homolog B, Scleraxis Homolog B (Mouse), SCXA, SCXB |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human SCX protein (Uniprot: Q7RTU7) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?