missing translation for 'onlineSavingsMsg'
Learn More

SCUBE3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18393635 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393635 100 μg 100µL
18368904 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18393635 Supplier Novus Biologicals Supplier No. NBP309503100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCUBE3 Polyclonal specifically detects SCUBE3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCUBE3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias CEGF3, CUB and EGF containing protein 3, CUB domain and EGF-like repeat containing 3, DKFZp686B09105, DKFZp686B1223, DKFZp686D20108, FLJ34743, signal peptide, CUB and EGF-like domain containing protein 3, signal peptide, CUB and EGF-like domain-containing protein 3, signal peptide, CUB domain, EGF-like 3
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human SCUBE3 (BAC03798). Peptide sequence MNCMNKNHGCAHICRETPKGGIACECRPGFELTKNQRDCKLTCNYGNGGC
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222663
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.