missing translation for 'onlineSavingsMsg'
Learn More

SCUBE2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18416563 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416563 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416563 Supplier Novus Biologicals Supplier No. NBP177186JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCUBE2 Polyclonal antibody specifically detects SCUBE2 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCUBE2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Cegb1, Cegf1, CEGP1, FLJ16792, FLJ35234, MGC133057, Protein CEGP1, signal peptide, CUB and EGF-like domain-containing protein 2, signal peptide, CUB domain, EGF-like 2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human SCUBE2. The immunogen is located within amino acids 480 - 530 of SCUBE2. Amino Acid Squence: CSPLRNKQQKSNDSAFGD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57758
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.