missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCUBE1 Polyclonal antibody specifically detects SCUBE1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | SCUBE1 |
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | signal peptide, CUB and EGF-like domain-containing protein 1, signal peptide, CUB domain, EGF-like 1, signal peptide-CUB domain-EGF-related 1 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 14 amino acid synthetic peptide near the center of human SCUBE1. The immunogen is located within amino acids 460 - 510 of SCUBE1. Amino Acid Squence: CSDAPTTPIKQKAR |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?