missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCTR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552411
This item is not returnable.
View return policy
Description
Immunogen sequence: CDVLQVLWEE QDQCLQELSR EQTGDLGTEQ PVPGCEGMWD NISCWPSSVP GRMVEVECPR FLRMLTSRNG SLFRNCTQDG WSETFPRPNL ACGVNVNDSS NEKRHSYLLK L Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 70%.
The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism.
Specifications
| SCTR | |
| Polyclonal | |
| Unconjugated | |
| Sctr | |
| 6530402O03Rik; MYOP; pancreatic secretin receptor; Sctr; SCT-R; Secretin receptor; SR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P47872 | |
| Sctr | |
| Recombinant protein corresponding to Human SCTR. Recombinant protein control fragment (Product #RP-88663). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction