missing translation for 'onlineSavingsMsg'
Learn More

BOLA Scrubber Bottles Vitrum

Product Code. 11328401
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11328401

Brand: BOLA N166214

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Scrubber Bottles Vitrum with two lateral GL 18 threaded necks bottle made of borosilicate glass 3.3

  • Consisting of PTFE body with screw cap and two lateral GL 18 threaded necks
  • Suitable bottle made of borosilicate glass as well as a FEP inlet tube and a gas distributor with finest bores.
  • Easy in- and outlet of gas by means of hard-walled tubing (e. g. PTFE) which can be connected to the threaded necks with BOLA Laboratory Screw Joints.
  • Elastic tubing can be connected by means of hose connectors.
  • The special feature: The body of the distributor can be turned independently from the screw cap.
  • The completely assembled distributor can be removed and fixed on another bottle without the risk of disarranging the tubing.

Specifications

Capacity (Metric) 500 mL
Height (Metric) 207 mm
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.