missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCRN2 Polyclonal antibody specifically detects SCRN2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | SCRN2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | secernin 2, secernin-2, Ses2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 176-425 of human SCRN2 (NP_612364.2).,, Sequence:, GRLWAAQRIQEGARNISNQLSIGTDISAQHPELRTHAQAKGWWDGQGAFDFAQIFSLTQQPVRMEAAKARFQAGRELLRQRQGGITAEVMMGILRDKESGICMDSGGFRTTASMVSVLPQDPTQPCVHFLTATPDPSRSVFKPFIFGMGVAQAPQVLSPTFGAQDPVRTLPRFQTQVDRRHTLYRGHQAALGLMERDQDRGQQLQQKQQDLEQEGLEATQGLLAGEWAPPLWELGSLFQAFVKRESQAYA |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?