missing translation for 'onlineSavingsMsg'
Learn More

SCRN2 Antibody, Novus Biologicals™

Product Code. 30230352 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30230352 100 μL 100µL
30230983 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30230352 Supplier Novus Biologicals Supplier No. NBP335207100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCRN2 Polyclonal antibody specifically detects SCRN2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCRN2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias secernin 2, secernin-2, Ses2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 176-425 of human SCRN2 (NP_612364.2).,, Sequence:, GRLWAAQRIQEGARNISNQLSIGTDISAQHPELRTHAQAKGWWDGQGAFDFAQIFSLTQQPVRMEAAKARFQAGRELLRQRQGGITAEVMMGILRDKESGICMDSGGFRTTASMVSVLPQDPTQPCVHFLTATPDPSRSVFKPFIFGMGVAQAPQVLSPTFGAQDPVRTLPRFQTQVDRRHTLYRGHQAALGLMERDQDRGQQLQQKQQDLEQEGLEATQGLLAGEWAPPLWELGSLFQAFVKRESQAYA
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 90507
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.