Learn More
Invitrogen™ SCRIB Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5116752
Product Code. 17239253
  • £454.00 / 100µL
Description
Positive Samples: A-549, 293T Immunogen sequence: ELCIQKAPGE RLGISIRGGA RGHAGNPRDP TDEGIFISKV SPTGAAGRDG RLRVGLRLLE VNQQSLLGLT HGEAVQLLRS VGDTLTVLVC DGFEASTDAA LEVSPGVIAN PFAAGIGHRN SLESISSIDR ELSPEGPGKE KELPGQTLHW GPEATEAAGR GLQPLKLDYR ALAAVPSAGS VQRVPSGAAG GKMAESPCSP SGQQPPSPPS PDELPANVKQ AYRAFAAVPT SHPPEDAPAQ PPTPGPAASP EQLSFRERQK YFELEVRVPQ AEGPPKRVSL VGADDLRKMQ EEEARKLQQK R.
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCRIB
Polyclonal
Unconjugated
SCRIB
AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul
Rabbit
Affinity chromatography
RUO
23513
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
1.19 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Q14160
SCRIB
Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1400 of human SCRIB (NP_8743653).