missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCRIB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554821
This item is not returnable.
View return policy
Description
Immunogen sequence: VSVIQFLEAP IGDEDAEEAA AEKRGLQRRA TPHPSELKVM KRSIEGRRSE ACPCQPDSGS PLPAEEEKRL SAESGLSEDS RPSASTVS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
SCRIB is a cytoplasmic multimodular scaffold protein targeted to epithelial adherens junctions and neuronal presynaptic compartments. SCRIB and its orthologs in vertebrates and invertebrates participate in cell polarization.
Specifications
| SCRIB | |
| Polyclonal | |
| Unconjugated | |
| SCRIB | |
| AI118201; circletail; Crc; CRIB; CRIB1; hScrib; Kiaa0147; LAP4; mKIAA0147; PDZ-domain protein scribble; Protein LAP4; Protein scribble homolog; RGD1565055; S Vartul; SCRB1; Scrib; Scrib1; Scribble; scribble homolog 1; scribbled homolog; scribbled homolog (Drosophila); scribbled planar cell polarity protein; Vartul | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23513 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.40 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14160 | |
| SCRIB | |
| Recombinant protein corresponding to Human SCRIB. Recombinant protein control fragment (Product #RP-93933). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction