Learn More
Globe Scientific Screwcap Sample Tubes, Non-Graduated
Product Code. 31923796 Shop All Globe Scientific Products
Change view
Click to view available options
Bottom Shape:
Conical (Self-standing)
Round
Round (Self-standing)
Capacity (Metric):
1.2 mL
2 mL
3 mL
4 mL
5 mL
Unit Size:
Pack of 1000
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bottom Shape Capacity (Metric) Unit Size
31923796 Round 4 mL Pack of 1000
31923641 Conical (Self-standing) 1.2 mL Pack of 1000
31921818 Round 2 mL Pack of 1000
31922475 Round (Self-standing) 2 mL Pack of 1000
31922964 Round (Self-standing) 3 mL Pack of 1000
31922344 Round (Self-standing) 4 mL Pack of 1000
31922265 Round (Self-standing) 5 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 31923796 Supplier Globe Scientific Supplier No. 6034
Estimated Shipment: 31-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
These tubes and caps have been designed for the storage and transportation of biological material, with non graduated
  • Tubes are manufactured from non-toxic polypropylene and offer strength and clarity with non-graduated
  • The vial has external threads, which provide a smooth and uniform inner surface which reduces the risk of contamination
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
  • The caps are available with or with out a silicone o-ring and are available in a variety of colors
  • The screw caps are also made of polypropylene, providing the same coefficient of expansion as the tubes
  • This along with a polypropylene inner lip ensures a leakproof seal, even at changing temperatures
  • The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn
Spezifikation
Autoclavable Autoclavable
Capacity (Metric) 4 mL
Bottom Shape Round
Material Polypropylene
Thread Style External
Sterility Non-sterile
Graduated No
Product Type Sample Tube
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.