missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Screw Caps with Multiple Distributors For Bottles

Product Code. 10787771
Change view
Click to view available options
Material:
PFA
PTFE
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10787771 PFA Each
10445461 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10787771 Supplier Bohlender™ Supplier No. D61408
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these screw caps with distributors for drawing or inserting aggressive or pure liquids. Bohlender™ BOLA™ Screw Caps with Multiple Distributors For Bottles are ideal for inserting tubing, tubes and probes into vessels.

  • Black screw cap made of PPS for bottle thread GL 45
  • Body made of PTFE or PFA
  • Insertion of tubing with a max. O.D. of 8.5 mm
  • Very good chemical resistance
  • Working temperature up to +200°C

Specifications

For Use With (Application) Drawing or inserting aggressive or pure liquids, Inserting tubing with max. 8.5 O.D., tubes and probes into vessels
Material PFA
Quantity 1Pack
Temperature Range (Metric) –200 to 250°C
Product Type Screw Cap
For Use With (Equipment) GL 45 thread
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.