missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Screw Caps for Microcentrifuge Tubes

Product Code. 11611300
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11611300

Brand: Simport™ Scientific T340NLSFT

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

These flat top caps accommodate automatic capping machines in packaging industries. The moulded internal lip fits snugly against the interior wall of the tube preventing the contents from coming in contact with the seal or threads, reducing the chances of sample contamination.

  • Moulded deep internal lip reduces sample contamination
  • High profile of cap facilitates manipulation and can remain attached to tube to eliminate mix-ups and contamination
  • Can be colour coded by the use of T345 Series Colored CapinsertTMwithout removing the cap
  • Flat top to accommodate automatic capping machines
  • Silicone washer seal
  • Polypropylene
  • Three cap styles available and two sealing types: washer seal and lip seal

Specifications

Includes Silicone washer seal
Liner Material Silicone Washer Seal
Product Type Screw Top
Material Polypropylene
Color Natural
For Use With (Application) The High Profile of This Flat Top Cap Can be Used in Automatic Capping Machines and Packaging Industries
For Use With (Equipment) Microcentrifuge Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.