missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S Murray™ Screw Cap Closure, 140 Pk.
Description
Screw Caps for use with T102/V3.
- Made of black polypropylene with foil-faced wads
Specifications
Specifications
| Liner Material | Foil |
| Material | Polypropylene |
| Color | Black |
| No. per Pack | 140 |
| For Use With (Equipment) | T102/V3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?