Learn More
Fisherbrand™ Screen
Product Code. 10589391
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand GD-5040 Gel Dryer System
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10589391 Fisherbrand GD-5040 Gel Dryer System Each
12358546 Fisherbrand GD-5040 Gel Dryer System Each
2 options
Product Code. 10589391
Brand: Fisherbrand™ VGDSSS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessory for use with vacuum gel dryers
  • Acts in conjunction with cast-aluminum base
  • Serve as porous gel support
  • Uniformly distributesthe heat and vacuum
  • Prevent gel cracking during drying
  • Pore size of stainless steel sheet: 1mm
Specifications
For Use With (Equipment) Fisherbrand GD-5040 Gel Dryer System
Type Screen