missing translation for 'onlineSavingsMsg'
Learn More

SCR-1481B1, MedChemExpress

Product Code. 16497516
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497516 100 mg 100mg
16467598 10 mg 10mg
16453617 50 mg 50mg
16400078 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497516 Supplier Medchem Express Supplier No. HY18711A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCR-1481B1 (c-Met inhibitor 2) is a potent compound that has activity against cancers dependent upon Met activation and also has activity against cancers as a VEGFR inhibitor.

SCR-1481B1 (c-Met inhibitor 2) is a potent compound that has activity against cancers dependent upon Met activation and also has activity against cancers as a VEGFR inhibitor.

Specifications

Chemical Name or Material SCR-1481B1
CAS 1174161-86-4
Color White
Purity Grade Notes Research
Molecular Formula C32H40ClF2N6O13P
Quantity 100 mg
Synonym c-Met inhibitor 2
Solubility Information DMSO : ≥ 100 mg/mL (142.86 mM)
SMILES O=C(C1=CN(COP(O)(O)=O)C=C(C2=CC=C(F)C=C2)C1=O)NC3=CC=C(OC4=C(Cl)C(N)=NC=C4)C(F)=C3.OCC(CO)(N)CO
Molecular Weight (g/mol) 699.98
Formula Weight 699.98
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.