missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP3/SYCP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 56 publications
£214.00 - £503.00
Specifications
| Dilution | Western Blot 0.5 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 18694876), Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:200-1:500, Immunoblotting reported in scientific literature (PMID 27612028) |
|---|---|
| Classification | Polyclonal |
| Target Species | Mouse, Rat, Pig, Chicken |
| Gene Alias | anti-SCP3, MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 |
| Isotype | IgG |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18728253
|
Novus Biologicals
NB300-232SS |
£214.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18751004
|
Novus Biologicals
NB300-232 |
£503.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
SCP3/SYCP3 Polyclonal specifically detects SCP3/SYCP3 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Feline, Parasite samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
| Western Blot 0.5 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 18694876), Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:200-1:500, Immunoblotting reported in scientific literature (PMID 27612028) | |
| Mouse, Rat, Pig, Chicken | |
| IgG | |
| Store at 4C. Do not freeze. |
| Polyclonal | |
| anti-SCP3, MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title