missing translation for 'onlineSavingsMsg'
Learn More

SCP3/SYCP3 Antibody (6F9C5) - Azide and BSA Free, Novus Biologicals™

Product Code. 18367550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367550 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367550 Supplier Novus Biologicals Supplier No. NBP280942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCP3/SYCP3 Monoclonal specifically detects SCP3/SYCP3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCP3/SYCP3
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 6F9C5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:500 - 1:2000, Flow Cytometry : 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry : 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence : 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready
Formulation PBS
Gene Alias MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3
Gene Symbols SYCP3
Host Species Mouse
Immunogen Partial recombinant human SCP3 (between residues 1-200) expressed in E. coli [Uniprot: Q8IZU3]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 50511
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.