missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558867
This item is not returnable.
View return policy
Description
Immunogen sequence: ILNMFRQQQK ILQQSRIVQS QRLKTIKQLY EQFIKSMEEL EKNHDNLLTG AQNEFKKEMA MLQKKIMMET QQQEIASVRK SLQSMLF Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregnancy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
| SCP3 | |
| Polyclonal | |
| Unconjugated | |
| Sycp3 | |
| Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50511 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZU3 | |
| Sycp3 | |
| Recombinant protein corresponding to Human SCP3. Recombinant protein control fragment (Product #RP-97796). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction