missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCP2 Recombinant Rabbit Monoclonal Antibody (24GB790)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifica
| Antigen | SCP2 |
|---|---|
| Clone | 24GB790 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
30275677
![]() |
Invitrogen™
MA557269 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274988
|
Invitrogen™
MA557270 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
SCP2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.Specifica
| SCP2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| SCP2 | |
| AA409774; AA409893; C76618; C79031; NLTP; nonspecific lipid transfer protein; Non-specific lipid-transfer protein; NSLIPTR; ns-LTP; NSL-TP; propanoyl-CoA C-acyltransferase; SCP2; SCP-2; SCP-CHI; SCPX; SCP-X; Sterol carrier protein 2; sterol carrier protein 2, liver; sterol carrier protein X | |
| SCP2 | |
| Primary | |
| Affinity chromatography |
| 24GB790 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11915, P22307, P32020 | |
| 20280, 25541, 6342 | |
| A synthesized peptide derived from human Sterol carrier protein 2 (aa 14-64). | |
| Antibody |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto