missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCP2 Monoclonal Antibody (OTI1E4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526335
This item is not returnable.
View return policy
Description
SCP2 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes two proteins: sterol carrier protein X (SCPx) and sterol carrier protein 2 (SCP2), as a result of transcription initiation from 2 independently regulated promoters. The transcript initiated from the proximal promoter encodes the longer SCPx protein, and the transcript initiated from the distal promoter encodes the shorter SCP2 protein, with the 2 proteins sharing a common C-terminus. Evidence suggests that the SCPx protein is a peroxisome-associated thiolase that is involved in the oxidation of branched chain fatty acids, while the SCP2 protein is thought to be an intracellular lipid transfer protein. This gene is highly expressed in organs involved in lipid metabolism, and may play a role in Zellweger syndrome, in which cells are deficient in peroxisomes and have impaired bile acid synthesis. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
Specifications
| SCP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22307 | |
| SCP2 | |
| Full length human recombinant protein of SCP2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E4 | |
| Unconjugated | |
| SCP2 | |
| AA409774; AA409893; C76618; C79031; NLTP; nonspecific lipid transfer protein; Non-specific lipid-transfer protein; NSLIPTR; ns-LTP; NSL-TP; propanoyl-CoA C-acyltransferase; SCP2; SCP-2; SCP-CHI; SCPX; SCP-X; Sterol carrier protein 2; sterol carrier protein 2, liver; sterol carrier protein X | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6342 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction